missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mark3 Antibody (OTI2G8), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72590DL594
This item is not returnable.
View return policy
Description
Mark3 Monoclonal antibody specifically detects Mark3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Mark3 | |
| Monoclonal | |
| DyLight 594 | |
| Cdc25C-associated protein kinase 1, cTAK1, C-TAK1, CTAK1ELKL motif kinase 2, EC 2.7.11, EC 2.7.11.1, EMK2, EMK-2, KP78, MAP/microtubule affinity-regulating kinase 3, PAR1A, Protein kinase STK10, Ser/Thr protein kinase PAR-1, Serine/threonine-protein kinase p78 | |
| Recombinant protein fragment corresponding to amino acids 473-729 of human Mark3 (NP_002367) produced in E.coli. | |
| 0.1 mL | |
| Protein Kinase | |
| 4140 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2G8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction