missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK3 Monoclonal Antibody (OTI3D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805884S
This item is not returnable.
View return policy
Description
MARK3 Monoclonal antibody specifically detects MARK3 in Human samples. It is validated for Western BlotSpecifications
| MARK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P27448 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4140 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3D8 | |
| Unconjugated | |
| MARK3 | |
| 1600015G02Rik; A430080F22Rik; cdc25C-associated protein kinase 1; cTAK1; C-TAK1; ELKL motif kinase 2; Emk2; EMK-2; EMK2/MARK3 serine-threonine protein kinase; ETK1; ETK-1; KP78; MAP/microtubule affinity regulating kinase 3; MAP/microtubule affinity-regulating kinase 3; MARK3; microtubule affinity regulating kinase 3; Mpk10; MPK-10; PAR1A; Par-1a; Protein kinase STK10; ser/Thr protein kinase PAR-1; Serine/threonine-protein kinase p78 | |
| Human recombinant protein fragment corresponding to amino acids 473-729 of human MARK3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction