missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARK4 Antibody (OTI9B7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72591PCP
This item is not returnable.
View return policy
Description
MARK4 Monoclonal antibody specifically detects MARK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MARK4 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI9B7 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.1, FLJ12177, FLJ90097, KIAA1860MARK4L, MAP/microtubule affinity-regulating kinase 4, MAP/microtubule affinity-regulating kinase like 1, MAP/microtubule affinity-regulating kinase-like 1, MARK4 serine/threonine protein kinase, MARKL1L, MARKL1MARK4S, Nbla00650 | |
| Human recombinant protein fragment corresponding to amino acids 390-467 of human MARK4(NP_113605) produced in E.coli. | |
| 0.1 mL | |
| Apoptosis | |
| 57787 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction