missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARVELD1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76243B
This item is not returnable.
View return policy
Description
MARVELD1 Polyclonal antibody specifically detects MARVELD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| MARVELD1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Biotin | |
| bA548K23.8, FLJ22343, FLJ23440, GB14, MARVD1, MARVEL (membrane-associating) domain containing 1, MARVEL domain containing 1, MGC4415, MRVLDC1, putative MARVEL domain-containing protein 1, RP11-548K23.8 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human MARVELD1. The immunogen is located within the first 50 amino acids of MARVELD1. Amino Acid Squence: PPPRQPPPQARAARGA | |
| 0.1 mL | |
| Cancer | |
| 83742 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction