missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARVELD2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74633AF532
This item is not returnable.
View return policy
Description
MARVELD2 Polyclonal antibody specifically detects MARVELD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| MARVELD2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| MARVEL domain containing 2, MRVLDC2, TRICautosomal recessive 49 | |
| The immunogen recognized by this antibody maps to a region between residue 90 and 140 of human MARVEL domain containing 2 (Tricellulin) using the numbering given in entry NP_001033692.1 (GeneID 153562). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 153562 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction