missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ CardioImmune™ XL Controls

Product Code. 15190027 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 mL
Level:
1
2
3
Level L
Unit Size:
18mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Level Unit Size
15190027 3 mL 3 18mL
15160027 3 mL Level L 18mL
15170027 3 mL 1 18mL
15180027 3 mL 2 18mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15190027 Supplier Thermo Scientific™ Supplier No. CAIXL3
  • ONLINE EXCLUSIVE PRICE
    € 304.00
    € 275.05 / 18mL
    Save € 28.95
    9% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls.

Monitor serum cardiac marker test methods with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ CardioImmune™ XL Controls. These controls are also available in a convenient Vista™ vial format.

  • Human serum matrix
  • Offers 4 distinctive levels
  • Assayed values available for most major analyzers
  • Available in a tri-level multipack
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 3 year shelf life at -25° to -15°C; 30 days open-vial at 2° to 8°C (for hsCRP, Digitoxin, Troponin I and Homocysteine); 15 days open-vial at 2° to 8°C (for BNP32, Myoglobin (when a dropper tip is used), proBNP, Troponin T, CK-MB)
Description CardioImmune XL Control, Assayed
Detectable Analytes Brain Natriuretic Peptide-32 (BNP-32), Myoglobin, Creatinine Kinase-MB (CK-MB), N-Terminal Brain Natriuretic Peptide (NT-proBNP), Digitoxin, Troponin I, Homocysteine, Troponin T, hcCRP
Sample Type Serum
Type Control
Level 3
Packaging 6 Pk.
Quantity 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.