missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAS™ Immunology Controls

Product Code. 18848398 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Immunology Controls, Assayed
Level:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Level
18848398 Immunology Controls, Assayed 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18848398 Supplier Thermo Scientific™ Supplier No. IC303
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte immunology controls.

Monitor the performance of various serum protein and serology test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ Immunology Controls. These controls are also available in a convenient Vista™ vial format.

  • Assayed for most major instrument systems
  • Three levels provide monitoring of the full assay range
  • Contains specific serum proteins, including Antistreptolysin, CRP, Prealbumin and Rheumatoid Factor
  • Human-serum matrix
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Two year shelf life from date of manufacture at 2° to 8°C; 30 day open vial stability for all analytes at 2° to 8°C
Description Immunology Controls, Assayed
Detectable Analytes Albumin, a-1-Acid Glycoprotein, a-1-Antitrypsin, a-2-Macroglobulin, Antistreptolysin, Antithrombin III, APO A (HDL Concentrate), APO B (LDL Concentrate), beta-2-microglobulin (B2M), C3 Complement, C4 Complement, Ceruloplasmin, Haptoglobin, IgA, IgE, IgG, IgM, Kappa Light Chain, Lambda Light Chain, Prealbumin, Properdin B, Rheumatoid Factor, Total Protein, Transferrin (TRF)
Sample Type Serum
Type Control
Level 3
Packaging 6 Pk.
Quantity 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.