missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAS1L Antibody (15B8), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | MAS1L |
|---|---|
| Clone | 15B8 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30031346
|
Novus Biologicals
NBP3-26148-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028992
|
Novus Biologicals
NBP3-26148-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAS1L Monoclonal antibody specifically detects MAS1L in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| MAS1L | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| dJ994E9.2, MAS1 oncogene-like, MAS1-like, MAS-L, MAS-R, mas-related G protein-coupled MRG, mas-related G-protein coupled receptor MRG, MRGMGC119987 | |
| A synthesized peptide derived from Human MAS1L [UniProt P35410] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 15B8 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 116511 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title