Learn More
MAS1L Antibody (SR1619), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-22564-50ul
 View more versions of this product
Product Code. 18604504
  • € 338.00 / 50µL
Description
MAS1L Monoclonal antibody specifically detects MAS1L in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAS1L
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunoprecipitation
SR1619
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
dJ994E9.2, MAS1 oncogene-like, MAS1-like, MAS-L, MAS-R, mas-related G protein-coupled MRG, mas-related G-protein coupled receptor MRG, MRGMGC119987
A synthesized peptide derived from human MAS1L (Uniprot #: P35410)
50 μg
GPCR
116511
Store at -20° C. Avoid freeze/thaw cycles.