missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAS1L Recombinant Rabbit Monoclonal Antibody (15B8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550060
This item is not returnable.
View return policy
Description
MAS1L Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
The proto-oncogene MAS1 is a G protein-coupled receptor located on the plasma membrane. In transfected NIH/3T3 cells, MAS1 has a weak focus-inducing activity. MAS1 is an antagonist of the AT1 receptor (Angiotensin II type 1 receptor), inhibiting the actions of Angiotensin II. MAS1L (MAS1-like), also known as MAS-L or MRG (Mas-related G-protein coupled receptor MRG), is a 378 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 (GPCR) family. MAS1 related proteins are thought to regulate nociceptor function and development, including the sensation or modulation of pain.
Specifications
| MAS1L | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35410, Q7TN38 | |
| MAS1L | |
| A synthesized peptide derived from human MAS1L. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 15B8 | |
| Unconjugated | |
| MAS1L | |
| DAAP-341G9.4; dJ994E9.2; MAS1 oncogene-like; MAS1 proto-oncogene like, G protein-coupled receptor; MAS1L; MAS1-like; MAS-L; MAS-R; mas-related G protein-coupled MRG; Mas-related G protein-coupled receptor b; MAS-related gene H; MAS-related GPR, member H; mas-related G-protein coupled receptor member H; Mas-related G-protein coupled receptor MRG; MAS-related G-protein coupled receptor, member H; MGC119987; Mgrb; MRG; Mrg and gpcr; Mrgh; MrgHGPR90; Mrgprh | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116511, 404641 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction