Learn More
Aalborg™ Mass Flow Controller for Nitrogen
Click to view available options
Certifications/Compliance:
NIST calibration certificate
Unit Size:
Each
Description
- Rigid metallic construction
- Built-in tiltable LCD readout
- Circuit protection
- Can be used as a portable device
- Engineering units or 0 to 100% displays
- TIO - Totalizer option
Specifications
Specifications
| Type | Mass Flowmeter |
| Accuracy | ±5% |
| Certifications/Compliance | NIST calibration certificate |
| Flow Rate Range | 0 to 5 LPM |
| Precision | ±1.5 % |
| Scale | Full scale |