Learn More
Invitrogen™ Mast Cell Chymase Recombinant Rabbit Monoclonal Antibody (23GB5095), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552788
 View more versions of this product
Product Code. 30088522
  • € 468.00 / 100µL
Description
This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11. 2 in a cluster of genes encoding other proteases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mast Cell Chymase
Recombinant Monoclonal
Unconjugated
CMA1
alpha-chymase; Chymase; chymase 1; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; CMA1; CYH; CYM; mast cell; Mast cell chymase 1; mast cell protease 3; Mast cell protease 5; mast cell protease I; Mast cell protease III; MCP3P; Mcp5; Mcp-5; Mcpt3; Mcpt5; MCT1; mMCP-5; rMCP-3; rMCP-5; rMCP-III
Rabbit
Affinity chromatography
RUO
17228
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5095
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P21844
CMA1
A synthesized peptide derived from human Mast Cell Chymase (204-247AA).