missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mast Cell Marker Antibody (MCG35), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50484F
This item is not returnable.
View return policy
Description
Mast Cell Marker Monoclonal specifically detects Mast Cell Marker in Human samples. It is validated for Immunohistochemistry.
Specifications
| Mast Cell Marker | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Spleen cells and bone marrow cells (erythrocyte depleted) from a patient with systemic mastocytosis. The bone marrow preparation consisted of 50% typical mast cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Immunohistochemistry | |
| MCG35 | |
| Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| Cardiovascular Biology | |
| The antibody does not react with basophils or other haemopoietic cell types. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction