missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mast Cell Protease-1/Mcpt1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98243AF350
This item is not returnable.
View return policy
Description
Mast Cell Protease-1/Mcpt1 Polyclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for ELISA.
Specifications
| Mast Cell Protease-1/Mcpt1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 17224 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 350 | |
| AV080368, Mcp-1 | |
| Produced in rabbits immunized with purified, recombinant Mast Cell Protease-1/Mcpt1 (NP_032596.1; Met 1-Lys 246). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction