Learn More
Mast Cell Protease-1/Mcpt1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98243AF750
Product Code. 18091948
  • ONLINE EXCLUSIVE PRICE
    € 438.00
    € 413.70 / 0.10mL
    Save € 24.30
    5% Off
Description
Mast Cell Protease-1/Mcpt1 Polyclonal specifically detects Mast Cell Protease-1/Mcpt1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mast Cell Protease-1/Mcpt1
Polyclonal
50 mM Sodium Borate
Rabbit
Affinity purified
RUO
17224
Store at 4°C in the dark.
IgG
ELISA
Alexa Fluor 750
AV080368, Mcp-1
Produced in rabbits immunized with purified, recombinant Mast Cell Protease-1/Mcpt1 (NP_032596.1; Met 1-Lys 246).
0.1 mL