missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MAST1 Antibody [mFluor Violet 500 SE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30749MFV500
This item is not returnable.
View return policy
Description
MAST1 Polyclonal antibody specifically detects MAST1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| MAST1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| EC 2.7.11, EC 2.7.11.1, KIAA0973microtubule-associated serine/threonine-protein kinase 1, microtubule associated serine/threonine kinase 1, SAST170, SASTsyntrophin associated serine/threonine kinase, Syntrophin-associated serine/threonine-protein kinase | |
| Produced in rabbits immunized with E. coli-derived Human MAST1 fragment | |
| 0.1 mL | |
| Protein Kinase | |
| 22983 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction