Learn More
Invitrogen™ MAST1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5114244
Product Code. 17132104
  • € 497.00 / 100µL
Description
Immunogen sequence: QESPLSLGAD PLLPEGASRP PVSSKEKESP GGAEACTPPR ATTPGGRTLE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Human: 100%; Pig: 100%.
Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MAST1
Polyclonal
Unconjugated
MAST1
9430008B02Rik; KIAA0973; MAST1; mast1 {ECO:0000312; microtubule associated serine/threonine kinase 1; microtubule-associated serine/threonine-protein kinase 1; RGD:631372}; SAST; SAST170; syntrophin associated serine/threonine kinase; syntrophin-associated serine/threonine-protein kinase; syntrophin-associated serine-threonine protein kinase
Rabbit
Affinity chromatography
RUO
22983
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y2H9
MAST1
Synthetic peptide directed towards the C-terminal region of Human MAST1.