missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAST2 Monoclonal Antibody (OTI1C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808791
This item is not returnable.
View return policy
Description
Syntrophin is an adapter protein that functions to bind certain signaling molecules to the dystrophin-associated protein complex. This complex connects the extracellular matrix to the intracellular cytoskeleton for construction and maintece of the postsynaptic structures in the neuromuscular junction and the central nervous system. Microtubule-associated serine/threonine-protein kinase 2 (MAST205) is a testis-specific, cytoplasmic protein that functions in a multi-protein complex in the maturation of spermatids. MAST205 is involved in linking the dystrophin/utrophin network with microtubule filaments via Syntrophin. By forming a complex with TRAF6, MAST205 regulates lipopolysaccharide-induced IL-12 synthesis in macrophages. This leads to the inhibition of TRAF6 NF-kappa-B activation. Two isoforms exist for MAST205 due to alternative splicing. Isoform 1 represents the full length protein, while isoform 2 lacks the residues 327-396 and 1091-1113. The N-terminus of MAST205 must be phosphorylated in order for ubiquitination to occur at the same site. This ubiquitination leads to the degradation of MAST205 via proteasome-mediated proteolysis.Specifications
| MAST2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P0Q8 | |
| MAST2 | |
| Human recombit protein fragment corresponding to amino acids 1482-1798 of human MAST2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1C12 | |
| Unconjugated | |
| MAST2 | |
| MAST205, MTSSK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23139 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction