missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAST2 Monoclonal Antibody (OTI1C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808791S
This item is not returnable.
View return policy
Description
MAST2 Monoclonal antibody specifically detects MAST2 in Human samples. It is validated for Western BlotSpecifications
| MAST2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P0Q8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23139 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1C12 | |
| Unconjugated | |
| MAST2 | |
| KIAA0807; Mast2; MAST205; microtubule associated serine/threonine kinase 2; microtubule associated testis specific serine/threonine protein kinase; microtubule-associated serine/threonine-protein kinase 2; Mtssk | |
| Human recombinant protein fragment corresponding to amino acids 1482-1798 of human MAST2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction