Learn More
Invitrogen™ MAST4 Monoclonal Antibody (K56010_15H12), Invitrogen™
Description
Sequence of this protein is as follows: KSAEPPRSPL LKRVQSEEKL SPSYGSDKKH LCSRKHSLEV TQEEVQREQS QREAPLQSLD ENVCDVPPLS RARPVEQGCL KRPVSRKVGR QESVDDLDRD KLKAKVVVKK ADGFPEKQES HQKSHGPGSD LENFALFKLE EREKKVYPKA VERSSTFENK ASMQEAPPLG SLLKDALHKQ ASVRASEGAM SDGRVPAEHR QGGGDFRRAP APGTLQDGLC HSLDRGISGK GEGTEKSSQA KELLRCEKLD SKLANIDYLR KKMSLEDKED NLCPVLKPKM TAGSHECLPG NPVRPTGGQQ EPPPASESRA
Specifications
Specifications
| Antigen | MAST4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | K56010_15H12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | MAST4 |
| Gene Accession No. | O15021 |
| Gene Alias | 4930420O11Rik; AI642422; KIAA0303; MAST4; microtubule associated serine/threonine kinase family member 4; microtubule-associated serine/threonine-protein kinase 4; mKIAA0303; RGD1310139 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.