Learn More
Greiner Bio-One MASTERBLOCK™ 96 Deep Well Conical Bottom 2mL Storage Plate
Product Code. 11397441
Click to view available options
Sterility:
Non-sterile
No. per Pack:
1
50
Unit Size:
Pack of 50
Product Code. 11397441
Brand: Greiner Bio-One 780280
Only null left
Add to basket
Add to basket
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Conical Bottom 2mL Storage Plate is also suitable for storing sample material.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
  • Square wells, Conical bottom
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 2 mL
Well Shape Conical Bottom (V-well)
No. per Case 50
No. per Pack 50
Quantity 50/Cs.
Well Design Square Wells