Learn More
Masterflex™ Handheld Remote Controllers
Masterflex™ Handheld Remote Controllers allow users to easily control Masterflex™ L/S™ pumps while filling and dispensing
Brand:  Masterflex™ 07528-80
Additional Details : Weight : 0.80000kg
Product Code. 12625646
  • € 4865.00 / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • Suitable for use with Masterflex™ L/S™ pumps
  • Long 10 ft cable makes remote control simple
  • Specifications
    Specifications
    One button
    Connection: DB9 (M)
    6 ft. (1.8 m) cable with noted terminal connector
    ABS