Learn More
Masterflex™ L/S™ End Bell Assembly
Complete your pump head assembly with two end bell assemblies. MasterFlex™ I/P™ End Bell Assemblies are designed for use with Masterflex™ I/P™ 73 Standard Pump Heads.
€ 145.00
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11885202
|
Masterflex™
07015-91 |
For Masterflex L/S Standard 07015-10/–21 pump heads |
€ 145.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
11895202
|
Masterflex™
07016-91 |
For Masterflex L/S Standard 07016-10/–21 pump heads |
€ 145.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Description
- Assemblies are made from high-performance Polyphenylene sulfide and stainless steel
Specifications
1 Each | |
End Bell Assembly |