Learn More
Masterflex™ PharmaPure™ I/P™ Precision Pump Tubing
Click to view available options
Tubing Size:
26
73
82
Unit Size:
Each
Description
Tubing is custom Extruded to fit Masterflex™ Pumps. Engineered for long life in peristaltic pump applications. Lot-to-lot consistency for superior accuracy and repeatability.
- Nontoxic and non-hemolytic with excellent biocompatibility
- Long life under pressure up to 60 psi (4 bar)
- Low extractables and low gas permeability
- Meets USP Class VI criteria
- FDA compliant
- Pharmaceutical biotech, and food applications
- Opaque off-white
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 9.5 mm |
| Material | PharmaPure |
| Overall Length (Metric) | 760 cm |
| For Use With (Application) | pharmaceutical, biotech and food applications |
| Min. Operating Temperature (Metric) | -51°C |
| Tubing Size | 73 |
| Flow Rate | 0.01 to 8L/min. |
| Height (Metric) | 8.8 m |
| Gas Permeability | Low |
| Show More |