missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT1A Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97484R
This item is not returnable.
View return policy
Description
MAT1A Polyclonal specifically detects MAT1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MAT1A | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AdoMet synthase 1, adoMet synthetase 1, AMS1, MAT, MATA1MAT 1, MAT-I/III, Methionine adenosyltransferase 1, methionine adenosyltransferase I, alpha, Methionine adenosyltransferase I/III, S-adenosylmethionine synthase isoform type-1, S-adenosylmethionine synthetase isoform type-1, SAMS, SAMS1EC 2.5.1.6 | |
| Produced in rabbits immunized with E. coli-derived Human MAT1A fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4143 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction