missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAT1A/MAT2A Recombinant Rabbit Monoclonal Antibody (HL1679)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547139
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
S adenosylmethionine synthetase catalyzes the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.
Specifications
| MAT1A/MAT2A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P31153, Q00266, Q3THS6, Q91X83 | |
| Mat1a, Mat2a | |
| Recombinant fragment of human MAT1A. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1679 | |
| Unconjugated | |
| Mat2a | |
| AdoMet; adoMet synthase 1; AdoMet synthase 2; adoMet synthetase 1; adoMet synthetase 2; AI046368; Ams; Ams1; Ams2; D630045P18Rik; MAT; MAT 1; MAT 2; Mat1a; Mat2a; MATA1; MATA2; MAT-I/III; MATII; MAT-II; Methionine adenosyltransferase 1; methionine adenosyltransferase 1A; methionine adenosyltransferase 2; methionine adenosyltransferase 2A; methionine adenosyltransferase I, alpha; methionine adenosyltransferase I/III; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S - adenosylmethionine synthetase; SADE; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-1; S-adenosylmethionine synthetase isoform type-2; SAMS; SAMS1; Sams2; SAS; testicular tissue protein Li 121 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11720, 232087, 4143, 4144 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction