missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAT1A Polyclonal Antibody
GREENER_CHOICE

Product Code. 17218333 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17218333 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17218333 Supplier Invitrogen™ Supplier No. PA5115549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAT1A.

MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and dominant mutations, the latter identified in autosomal dominant persistant hypermethioninemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAT1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MAT1A
Gene Accession No. P13444, Q00266, Q91X83
Gene Alias AdoMet; AdoMet synthase 1; adoMet synthetase 1; AI046368; Ams; Ams1; Cartilage matrix protein; CMP; CRTM; MAT; MAT 1; Mat1a; MATA1; MAT-I/III; MATN1; matrilin 1, cartilage matrix protein; Matrilin-1; Methionine adenosyltransferase 1; methionine adenosyltransferase 1A; methionine adenosyltransferase I, alpha; methionine adenosyltransferase I/III; S - adenosylmethionine synthetase; SADE; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase isoform type-1; S-adenosylmethionine synthase; S-adenosylmethionine synthetase isoform type-1; SAMS; SAMS1; SAS; similar to S-adenosylmethionine synthetase alpha and beta forms (Methionine adenosyltransferase) (AdoMet synthetase) (MAT-I/III); wu:fi35e01; zgc:55442
Gene Symbols MAT1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAT1A(Accession Q00266), corresponding to amino acid residues M1-I33.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11720, 25331, 4143
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.