missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT2A Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-94158AF532
This item is not returnable.
View return policy
Description
MAT2A Polyclonal specifically detects MAT2A in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MAT2A | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
| Rabbit | |
| 0.1 mL | |
| 4144 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MAT2A | |
| Synthetic peptide made to an N-terminal portion of human MAT2A (within residues 1-100). [Swiss-ProtNo. P31153] | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction