Learn More
MAT2A Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32283AF594
This item is not returnable.
View return policy
Description
MAT2A Polyclonal antibody specifically detects MAT2A in Human, Mouse samples. It is validated for Western Blot