missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAT2A Recombinant Rabbit Monoclonal Antibody (7A0X1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537899
This item is not returnable.
View return policy
Description
Positive Samples: HepG2, Mouse liver, Rat liver Immunogen sequence: MNGQLNGFHE AFIEEGTFLF TSESVGEGHP DKICDQISDA VLDAHLQQDP DAKVACETVA KTGMILLAGE ITSRAAVDYQ KVVREAVKHI GYDDSSKGFD.
The protein encoded by this gene catalyzes the production of S-adenosylmethionine (AdoMet) from methionine and ATP. AdoMet is the key methyl donor in cellular processes.
Specifications
| MAT2A | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18298, P31153, Q3THS6 | |
| MAT2A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MAT2A (P31153). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7A0X1 | |
| Unconjugated | |
| MAT2A | |
| adoMet synthase 2; adoMet synthetase 2; AMS2; D630045P18Rik; MAT 2; MAT2A; MATA2; MATII; MAT-II; Methionine adenosyltransferase 2; methionine adenosyltransferase 2A; Methionine adenosyltransferase II; methionine adenosyltransferase II, alpha; non-hepatic-type S-adenosylmethionine synthetase; S-adenosylmethionine synthase; S-adenosylmethionine synthase isoform type-2; S-adenosylmethionine synthetase isoform type-2; Sams2; testicular tissue protein Li 121 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171347, 232087, 4144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction