missing translation for 'onlineSavingsMsg'
Learn More

Cadence Science Dissolved gas analysis 50mL in 2mL, Lock Tip

Brand:  Cadence Science 5157

Dissolved gas analysis 50mL in 2mL, Lock Tip

Additional Details : Weight : 0.18000kg

Product Code. 15748347

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.