missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATOH1 Polyclonal Antibody

MATH1 Polyclonal Antibody
Brand: Invitrogen PA597480
This item is not returnable.
View return policy
Description
MATH1 Polyclonal Antibody for Western Blot, ELISA
Specifications
| ATOH1 | |
| Polyclonal | |
| Unconjugated | |
| Atoh1 | |
| ATH1; Atoh1; atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog 1 (Drosophila); atonal homolog bHLH transcription factor 1; BHLHA14; Class A basic helix-loop-helix protein 14; H MATH-1; Hath1; Helix-loop-helix protein hATH-1; helix-loop-helix protein mATH-1; Math1; MATH-1; Protein atonal homolog 1; RGD1565171 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11921 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P48985 | |
| Atoh1 | |
| Recombinant Mouse Protein atonal homolog 1 protein (1-351AA). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction