Learn More
Invitrogen™ MATK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110689
Description
Immunogen sequence: GTQCITKCEH TRPKPGELAF RKGDVVTILE ACENKSWYRV KHHTSGQEGL LAAGALRERE ALSADPKLSL MPWFHGKISG QEAVQQLQPP EDGLFLVRES ARHPGDYVLC VSFGRDVIHY RVLHRDGHLT IDEAVFFCNL MDMVEHY Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 92%.
MATK (Hyl) is a non-receptor CSK-type protein tyrosine kinase that is thought to function in signal transduction pathways important in megakaryocyte growth and/or differentiation. This protein is thought to play a significant role in the signal transduction of hematopoietic cells. It is able to phosphorylate and inactivate Src family kinases, and may play an inhibitory role in the control of T-cell proliferation. This protein might be involved in signaling in some cases of breast cancer.
Specifications
| MATK | |
| Polyclonal | |
| Unconjugated | |
| MATK | |
| Batk; CHK; Csk homologous kinase; Csk-homologous kinase; Csk-type protein tyrosine kinase; CTK; DKFZp434N1212; Hematopoietic consensus tyrosine-lacking kinase; HHYLTK; hydroxyaryl-protein kinase; HYL; HYL tyrosine kinase; HYLTK; leukocyte carboxyl-terminal src kinase related; Lsk; Matk; megakaryocyte-associated tyrosine kinase; megakaryocyte-associated tyrosine kinase (non-receptor protein tyrosine kinase); megakaryocyte-associated tyrosine-protein kinase; MGC1708; MGC2101; non-receptor protein kinase protein; Ntk; Protein kinase BATK; protein kinase HYL; Protein kinase NTK; tyrosine kinase MATK; tyrosine-protein kinase CTK; tyrosylprotein kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P42679 | |
| MATK | |
| Recombinant Protein corresonding to Human MATK. Recombinant protein control fragment (Product #RP-102111). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.