missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATN3 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77022V
This item is not returnable.
View return policy
Description
MATN3 Polyclonal antibody specifically detects MATN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| MATN3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 405 | |
| DIPOA, EDM5, HOA, matrilin 3, matrilin-3, OADIP, OS2 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the center of human MATN3. The immunogen is located within amino acids 180 - 230 of MATN3. Amino Acid Squence: VTDGRPQDQVNEV | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 4148 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction