Learn More
Invitrogen™ MATR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA541885
Product Code. 15565102
  • € 520.00 / 100µL
Description
Peptide sequence: MSKSFQQSSL SRDSQGHGRD LSAAGIGLLA AATQSLSMPA SLGRMNQGTA Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
This gene encodes a nuclear matrix protein, which is proposed to stabilize certain messenger RNA species. Mutations of this gene are associated with distal myopathy 2, which often includes vocal cord and pharyngeal weakness. Alternatively spliced transcript variants, including read-through transcripts composed of the upstream small nucleolar RNA host gene 4 (non-protein coding) and matrin 3 gene sequence, have been identified. Pseudogenes of this gene are located on chromosomes 1 and X.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MATR3
Polyclonal
Unconjugated
MATR3
1110061A14Rik; 2810017I02Rik; AI841759; ALS21; AW555618; D030046F20Rik; KIAA0723; MATR3; matrin 3; matrin-3; matrin-3-like protein; mKIAA0723; MPD2; nuclear scaffold protein P130/MAT3; P130/MAT3; VCPDM; vocal cord and pharyngeal weakness with distal myopathy
Rabbit
Protein A
RUO
9782
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P43243
MATR3
Synthetic peptide directed towards the N-terminal of human MATR3 (aa 1-50).