Learn More
Mavoglurant, TRC
Specifications
Specifications
| Chemical Name or Material | Mavoglurant |
| CAS | 543906-09-8 |
| Molecular Formula | C19H23NO3 |
| Quantity | 25 mg |
| Synonym | (3aR,4S,7aR)-Octahydro-4-hydroxy-4-[(3-methylphenyl)ethynyl]-1H-indole-1-carboxylic Acid Methyl Ester, AFQ 056 |
| InChI Formula | InChI=1S/C19H23NO3/c1-14-5-3-6-15(13-14)8-11-19(22)10-4-7-17-16(19)9-12-20(17)18(21)23-2/h3,5-6,13,16-17,22H,4,7,9-10,12H2,1-2H3/t16-,17-,19-/m1/s1 |
| SMILES | O=C(N1CC[C@]2([H])[C@@]1([H])CCC[C@]2(C#CC3=CC=CC(C)=C3)O)OC |
| IUPAC Name | Methyl (3aR,4S,7aR)-4-Hydroxy-4-(m-tolylethynyl)octahydro-1H-indole-1-carboxylate |
| Molecular Weight (g/mol) | 313.39 |
| Formula Weight | 313.17 |