missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS Antibody (58N3B6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22380F
This item is not returnable.
View return policy
Description
MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MAVS | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MAVS | |
| Amino acids 93-276 of human MAVS (isoform 1) were used as the immunogen for this antibody. | |
| 0.1 mL | |
| Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase | |
| 57506 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 58N3B6 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction