missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS Antibody (981220), DyLight 550, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB96941R
This item is not returnable.
View return policy
Description
MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MAVS | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAVS | |
| E.coli-derived partial recombinant human MAVS protein (amino acids 37-513) | |
| 0.1 mL | |
| Immune Dysfunction, Immune System Diseases, Immunology, Protein Kinase | |
| Human | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence | |
| 981220 | |
| Immunocytochemistry/Immunofluorescence | |
| CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISAFLJ38051 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction