Learn More
MAVS VISA Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-14341-100UL
Product Code. 16860566
  • € 450.00 / 100µL
Description
The MAVS; VISA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
MAVS VISA
Polyclonal
CoraLite 488
100 μL
57506
IgG
Immunofluorescence
1000 μg/mL