Learn More
MAVS VISA Rabbit, anti-Human, CoraLite Plus 488, Clone: 7F14, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-81910-100UL
Description
The MAVS; VISA antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MAVS VISA | |
| 7F14 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, FLJ35386, FLJ38051, FLJ41962, IPS 1, IPS1, KIAA1271, MAVS, VISA | |
| 100 μL | |
| 57506 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |