missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Max Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98815FR
This item is not returnable.
View return policy
Description
Max Polyclonal antibody specifically detects Max in Human samples. It is validated for Western Blot
Specifications
| Max | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| BHLHD4, bHLHd4MGC11225, bHLHd5, bHLHd6, bHLHd7, bHLHd8, Class D basic helix-loop-helix protein 4, helix-loop-helix zipper protein, MAX protein, MGC10775, MGC18164, MGC34679, MGC36767, MYC associated factor X, Myc-associated factor X, orf1, protein max | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Max. | |
| 0.1 mL | |
| Apoptosis | |
| 4149 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction