Learn More
Emma Safety Footwear Max MF Safety Shoes
Description
- S3-Safety shoe with sole plate for wet work
- Microfber construction
- TPU GripForce™ Easy Twist Outsole
- Metal-free
- Chrome-free lining
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black |
| Design | Max MF |
| Liner Material | Hydro-Tec™ Antibacterial |
| Material | Microfiber |
| Size | 40 |
| Toe Material | Composite |