Learn More
Tocris Bioscience™ MaxiPost
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MaxiPost is a potassium channel modulator. Acts as a positive modulator at neuronal Kv7 channels and calcium-activated K+ channels (BKCa) in HEK293 cells. Displays negative modulatory activity at Kv7.1 channels (Ki = 3.7 μM) and GABAA receptors. Displays anxiolytic activity in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | MaxiPost |
| Purity | >98% |
| Molecular Weight (g/mol) | 359.7 |
| Inhibitors | MaxiPost |