missing translation for 'onlineSavingsMsg'
Learn More

MB05032, MedChemExpress

Product Code. 16499567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499567 5 mg 5mg
16453077 50 mg 50mg
16447086 100 mg 100mg
16437028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499567 Supplier Medchem Express Supplier No. HY16307

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MB05032 is a special and efficacious gluconeogenesis inhibitor targeted the AMP binding site of fructose 1,6-bisphosphatase (FBPase) with an IC50 value of 16 nM.

MB05032 is a special and efficacious gluconeogenesis inhibitor targeted the AMP binding site of fructose 1,6-bisphosphatase (FBPase) with an IC50 value of 16 nM.

Specifications

Chemical Name or Material MB05032
Physical Form Solid
CAS 261365-11-1
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H15N2O4PS
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 50 mg/mL (165.40 mM; Need ultrasonic)
SMILES CC(C)CC1=C(C2=CC=C(P(O)(O)=O)O2)N=C(N)S1
Molecular Weight (g/mol) 302.29
Formula Weight 302.29
Percent Purity 99.69%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.