missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD1 Antibody (100B272.1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56537R
This item is not returnable.
View return policy
Description
MBD1 Monoclonal specifically detects MBD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications
| MBD1 | |
| Monoclonal | |
| DyLight 550 | |
| Q9UIS9 | |
| MBD1 | |
| This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1; GenBank no ref NP_056671.2 | |
| 0.1 mL | |
| Chromatin Research | |
| 4152 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), KnockDown | |
| 100B272.1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockdown Validated | |
| CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| There are many isoforms of human MBD1 ranging from 503 (∼55 kDa) to 655 (∼71 kDa) amino acids in length. Researchers are encouraged to use the provided immunogen sequence to determine if this antibody is suitable for testing a specific isoform of interest. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction