Learn More
UCH-L3 Antibody (204), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90680V
Product Code. 18262126
  • € 617.00 / 0.10mL
Description
UCH-L3 Monoclonal specifically detects UCH-L3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UCH-L3
Monoclonal
DyLight 405
50mM Sodium Borate
Rabbit
Protein A purified
RUO
7347
Mouse
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
204
Immunohistochemistry, Immunohistochemistry-Paraffin
EC 3.4.19.12, ubiquitin carboxyl-terminal esterase L3 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L3, Ubiquitin thioesterase L3, ubiquitin thiolesterase, UCH-L3
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse UCH-L3 (Q9JKB1; Glu2-Ala230).
0.1 mL
Primary
No cross-reactivity in ELISA with: Human UCHL3
Store at 4°C in the dark.