missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | MBD3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696688
|
Novus Biologicals
NBP3-00136-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647107
|
Novus Biologicals
NBP3-00136-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBD3 Polyclonal antibody specifically detects MBD3 in Human samples. It is validated for Western Blot, ELISASpecifications
| MBD3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| 0.2 um filtered solution in PBS | |
| 53615 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3 | |
| Produced in rabbits immunized with purified, recombinant Human MBD3 (O95983-2; Met 1-Val 259) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title