missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21219-100ul
This item is not returnable.
View return policy
Description
MBD4 Polyclonal antibody specifically detects MBD4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MBD4 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LQNQSNNSNWNLRTRSKCKKDVFMPPSSSSELQESRGLSNFTSTHLLLKEDEGVDDVNFRKVRKPKGKVTILKGIPIKKTKKGCRKSCSGFVQSDSKRESVCNKADAESEPVAQKSQLDRTVCISDAGACGETLSVTSEEN | |
| 100 μg | |
| Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics | |
| 8930 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction