missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBD5 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99368CL1
This item is not returnable.
View return policy
Description
MBD5 Polyclonal antibody specifically detects MBD5 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| MBD5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55777 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| KIAA1461, methyl-CpG binding domain protein 5, methyl-CpG-binding domain protein 5, methyl-CpG-binding protein MBD5, MRD1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MBD5. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction