missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MBD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565163
This item is not returnable.
View return policy
Description
Immunogen sequence: PGGGTNATPVV PSRAATPRSV RNKSHEGITN SVMPECKNPF KLMIGSSNAM GRLYVQELPG SQQQELHPVY PRQRLGSSEH GQKSPFRGSH Highest antigen sequence identity to the following orthologs - mouse 97%, rat 89%.
MBD5 binds to heterochromatin but does not interact with either methylated or unmethylated DNA (in vitro).
Specifications
| MBD5 | |
| Polyclonal | |
| Unconjugated | |
| MBD5 | |
| 9430004D19Rik; AA536666; AI426407; C030040A15Rik; FLJ11113; FLJ30517; Gm1630; KIAA1461; MBD5; methyl-CpG binding domain protein 5; methyl-CpG-binding domain protein 5; Methyl-CpG-binding protein MBD5; MRD1; OTTMUSG00000012483 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55777 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P267 | |
| MBD5 | |
| Recombinant Human MBD5. Recombinant protein control fragment (Product #RP-105914). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction